siRNA Details
| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023197.3 | NP_075686.2 | 2 | 846 | ||
| GenBank mRNA | AF034580.1 | - | 3 | 1013 | ||
| AK166956.1 | - | 2 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Knop1| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023197.3 | NP_075686.2 | 2 | 846 | ||
| GenBank mRNA | AF034580.1 | - | 3 | 1013 | ||
| AK166956.1 | - | 2 | 532 | |||