siRNA Details
| Gene Symbol: Eef1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285429.1 | NP_001272358.1 | 7 | 751 | ||
| NM_001285430.1 | NP_001272359.1 | 7 | 751 | |||
| NM_001285431.1 | NP_001272360.1 | 6 | 679 | |||
| NM_001285432.1 | NP_001272361.1 | 6 | 682 | |||
| NM_001285433.1 | NP_001272362.1 | 6 | 679 | |||
| NM_001285434.1 | NP_001272363.1 | 7 | 736 | |||
| NM_023240.3 | NP_075729.2 | 7 | 754 | |||
| NM_029663.2 | NP_083939.1 | 9 | 1899 | |||
| GenBank mRNA | AF304351.1 | - | 7 | 717 | ||
| AK010308.1 | - | 3 | 370 | |||
| AK010347.1 | - | 8 | 729 | |||
| AK017616.1 | - | 6 | 634 | |||
| AK017796.1 | - | 7 | 715 | |||
| AK044934.1 | - | 7 | 706 | |||
| BC013059.1 | - | 7 | 684 | |||
| BC079855.1 | - | 9 | 1845 | 6403006 | ||
| BC082536.1 | - | 9 | 1830 | |||
| BQ177630.1 | - | 7 | 720 | |||
| DV062370.1 | - | 6 | 686 | |||

Eef1d
Genomic Map