siRNA Details
| Gene Symbol: Ndufaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023247.1 | NP_075736.1 | 2 | 547 | ||
| GenBank mRNA | AF235017.1 | - | 2 | 547 | ||
| AK005252.1 | - | 2 | 162 | |||
| AK014634.1 | - | 2 | 162 | |||
| BC027513.1 | - | 2 | 580 | 5118121 | ||

Ndufaf3
Genomic Map