siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023270.5 | NP_075759.3 | 1 | 523 | ||
| GenBank mRNA | AB041548.1 | - | 1 | 523 | ||
| AF426411.1 | - | 1 | 337 | |||
| AK004847.1 | - | 1 | 524 | |||
| AK008312.1 | - | 1 | 467 | |||
| AK018582.1 | - | 1 | 525 | |||
| AK146266.1 | - | 1 | 520 | |||
| AK167031.1 | - | 1 | 509 | |||
| AY112656.1 | - | 1 | 510 | |||
| BC010477.1 | - | 1 | 512 | 3154433 | ||

Rnf128
Genomic Map