siRNA Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290454.1 | NP_001277383.1 | 4 | 1033 | ||
| NM_023305.3 | NP_075794.3 | 4 | 1033 | |||
| GenBank mRNA | AF275549.3 | - | 4 | 992 | ||
| AK009596.1 | - | 4 | 992 | |||
| AK028477.1 | - | 4 | 992 | |||
| AK028670.1 | - | 4 | 984 | |||
| AK030370.1 | - | 4 | 974 | |||
| AK033121.1 | - | 4 | 985 | |||
| AK051346.1 | - | 214 | ||||
| AK076078.1 | - | 4 | 992 | |||
| AK163229.1 | - | 4 | 991 | |||
| BC065996.1 | - | 4 | 981 | 5689710 | ||

Ubap1
Genomic Map