siRNA Details
| Gene Symbol: Ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141930.1 | NP_001135402.1 | 4 | 1285 | ||
| NM_023465.4 | NP_075954.1 | 5 | 1391 | |||
| GenBank mRNA | AB021261.1 | - | 5 | 1309 | ||
| AK036181.1 | - | 5 | 1394 | |||
| AK048258.1 | - | 5 | 1389 | |||
| BQ043263.1 | - | 1 | 654 | |||

Ctnnbip1
Genomic Map