siRNA Details
| Gene Symbol: Asb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023906.3 | NP_076395.2 | 5 | 774 | ||
| GenBank mRNA | AF155354.1 | - | 4 | 589 | ||
| AK010315.1 | - | 5 | 710 | |||
| AK028489.1 | - | 6 | 880 | |||
| AK038086.1 | - | 5 | 755 | |||
| AK143580.1 | - | 5 | 709 | |||
| BC023086.1 | - | 5 | 708 | 4947610 | ||

Asb3
Genomic Map