siRNA Details
| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024181.2 | NP_077143.2 | 18, 19 | 2425 | ||
| GenBank mRNA | AF255459.1 | - | 18, 19 | 2102 | ||
| AF314002.1 | - | 17, 18 | 1938 | |||
| AK004617.1 | - | 18, 19 | 2180 | |||
| AK028184.1 | - | 18, 19 | 2166 | |||
| AK128945.1 | - | 18, 19 | 2179 | |||
| AK165652.1 | - | 18, 19 | 2182 | |||
| AK166502.1 | - | 18, 19 | 2136 | |||
| AK169020.1 | - | 18, 19 | 2192 | |||
| BC002207.1 | - | 9, 10 | 987 | |||
| BC033461.1 | - | 18, 19 | 2164 | 4010341 | ||

Dnajc10
Genomic Map