siRNA Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024182.4 | NP_077144.2 | 3 | 354 | ||
| GenBank mRNA | AK004748.1 | - | 3 | 446 | ||
| AK131635.1 | - | 3 | 358 | |||
| AK134327.1 | - | 3 | 446 | |||
| AK147074.1 | - | 3 | 355 | |||
| AK167309.1 | - | 3 | 450 | |||
| AK170882.1 | - | 3 | 434 | |||
| BC033271.1 | - | 3 | 392 | 4016197 | ||

Riok3
Genomic Map