siRNA Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024182.4 | NP_077144.2 | 6 | 669 | ||
| GenBank mRNA | AK004748.1 | - | 6 | 761 | ||
| AK131635.1 | - | 6 | 673 | |||
| AK134327.1 | - | 6 | 761 | |||
| AK147074.1 | - | 6 | 670 | |||
| AK167309.1 | - | 6 | 765 | |||
| AK170882.1 | - | 6 | 749 | |||
| BC002255.1 | - | 3 | 199 | |||
| BC033271.1 | - | 6 | 707 | 4016197 | ||

Riok3
Genomic Map