siRNA Details
| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024184.2 | NP_077146.1 | 4 | 1166 | ||
| GenBank mRNA | AK005646.1 | - | 4 | 1199 | ||
| AK077005.1 | - | 4 | 1192 | |||
| AK077718.1 | - | 4 | 1147 | |||
| AK134906.1 | - | 4 | 1354 | |||
| AK150818.1 | - | 4 | 1147 | |||
| AK151951.1 | - | 4 | 1147 | |||
| AK165322.1 | - | 4 | 1145 | |||
| BC003428.1 | - | 4 | 1166 | 2645378 | ||

Asf1b
Genomic Map