siRNA Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024185.4 | NP_077147.2 | 4 | 465 | ||
| GenBank mRNA | AK002900.1 | - | 4 | 416 | ||
| AK009891.1 | - | 4 | 444 | |||
| AK010050.1 | - | 4 | 441 | |||
| AK049694.1 | - | 4 | 447 | |||
| AK049869.1 | - | 4 | 441 | |||
| AK143250.1 | - | 4 | 437 | |||
| BC002225.1 | - | 4 | 447 | |||
| BC027202.1 | - | 4 | 369 | 4217391 | ||

Fam188a
Genomic Map