siRNA Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024188.6 | NP_077150.1 | 15 | 1753 | ||
| GenBank mRNA | AB085609.1 | - | 15 | 1495 | ||
| AK009027.1 | - | 15 | 1533 | |||
| AK010029.1 | - | 2 | 136 | |||
| AK011354.1 | - | 15 | 1433 | |||
| AK078086.1 | - | 15 | 1751 | |||
| AK146158.1 | - | 15 | 1511 | |||
| AK146342.1 | - | 15 | 1498 | |||
| AK151662.1 | - | 15 | 1492 | |||
| AK151695.1 | - | 15 | 1483 | |||
| AK167681.1 | - | 15 | 1583 | |||
| BC003422.1 | - | 15 | 1457 | 2598824 | ||

Oxct1
Genomic Map