siRNA Details
| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025316.2 | NP_079592.2 | 5, 6 | 465 | ||
| GenBank mRNA | AK002279.1 | - | 5, 6 | 467 | ||
| AK004372.1 | - | 5, 6 | 465 | |||
| AK005521.1 | - | 5, 6 | 465 | |||
| AK013223.1 | - | 5, 6 | 467 | |||
| AK018562.1 | - | 5, 6 | 472 | |||
| AK088061.1 | - | 5, 6 | 474 | |||
| BC025155.1 | - | 5, 6 | 458 | 4024605 | ||

Ndufb5
Genomic Map