siRNA Details
| Gene Symbol: 0610009B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025319.2 | NP_079595.1 | 2 | 638 | ||
| GenBank mRNA | AK002369.1 | - | 2 | 638 | ||
| AK004363.1 | - | 2 | 637 | |||
| BC024353.1 | - | 2 | 604 | 5009365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610009B22Rik| Gene Symbol: 0610009B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025319.2 | NP_079595.1 | 2 | 638 | ||
| GenBank mRNA | AK002369.1 | - | 2 | 638 | ||
| AK004363.1 | - | 2 | 637 | |||
| BC024353.1 | - | 2 | 604 | 5009365 | ||