siRNA Details
| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025323.2 | NP_079599.1 | 4 | 812 | ||
| GenBank mRNA | BC019535.1 | - | 4 | 812 | 4458983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3b6| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025323.2 | NP_079599.1 | 4 | 812 | ||
| GenBank mRNA | BC019535.1 | - | 4 | 812 | 4458983 | |