siRNA Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025329.3 | NP_079605.1 | 2 | 242 | ||
| GenBank mRNA | AK002589.1 | - | 2 | 201 | ||
| AK004191.1 | - | 2 | 170 | |||
| AK007178.1 | - | 2 | 230 | |||
| AK010027.1 | - | 2 | 200 | |||
| AK010464.1 | - | 2 | 198 | |||
| BC059716.1 | - | 2 | 243 | 6774264 | ||

Tctex1d2
Genomic Map