siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025332.3 | NP_079608.1 | 1 | 163 | ||
| GenBank mRNA | AK002771.1 | - | 1 | 148 | ||
| AK010979.1 | - | 1 | 160 | |||
| AK078242.1 | - | 1 | 164 | |||
| AK147840.1 | - | 1 | 147 | |||
| BC107216.1 | - | 1 | 160 | 40051675 | ||
| CJ169587.1 | - | 1 | 164 | |||

Gtpbp8
Genomic Map