siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159329.1 | NP_001152801.1 | 3 | 335 | ||
| NM_025332.3 | NP_079608.1 | 3 | 536 | |||
| GenBank mRNA | AK002771.1 | - | 3 | 521 | ||
| AK010979.1 | - | 3 | 533 | |||
| AK147840.1 | - | 3 | 520 | |||
| BC107216.1 | - | 3 | 533 | 40051675 | ||
| BC107217.1 | - | 3 | 332 | 40051680 | ||

Gtpbp8
Genomic Map