siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159329.1 | NP_001152801.1 | 6 | 796 | ||
| NM_025332.3 | NP_079608.1 | 6 | 997 | |||
| GenBank mRNA | AK010979.1 | - | 6 | 994 | ||
| AK147840.1 | - | 6 | 981 | |||
| BC107216.1 | - | 6 | 994 | 40051675 | ||
| BC107217.1 | - | 6 | 793 | 40051680 | ||

Gtpbp8
Genomic Map