siRNA Details
| Gene Symbol: Tmem167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025335.3 | NP_079611.2 | 4 | 641 | ||
| GenBank mRNA | AK002884.1 | - | 4 | 567 | ||
| AK007966.1 | - | 4 | 575 | |||
| AK012161.1 | - | 4 | 575 | |||
| AK016036.1 | - | 4 | 576 | |||
| AK016737.1 | - | 1186 | ||||
| AK021263.1 | - | 4 | 587 | |||
| AK029487.1 | - | 1 | 329 | |||
| AK031706.1 | - | 4 | 576 | |||
| AK032474.1 | - | 4 | 585 | |||
| AK088895.1 | - | 4 | 563 | |||
| AK132361.1 | - | 4 | 563 | |||
| AK161037.1 | - | 4 | 578 | |||
| BC085489.1 | - | 4 | 548 | |||

Tmem167
Genomic Map