siRNA Details
| Gene Symbol: Tmem42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025339.4 | NP_079615.1 | 3 | 592 | ||
| GenBank mRNA | AK002688.1 | - | 3 | 568 | ||
| AK012820.1 | - | 3 | 567 | |||
| BC049583.1 | - | 3 | 592 | 6742203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem42| Gene Symbol: Tmem42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025339.4 | NP_079615.1 | 3 | 592 | ||
| GenBank mRNA | AK002688.1 | - | 3 | 568 | ||
| AK012820.1 | - | 3 | 567 | |||
| BC049583.1 | - | 3 | 592 | 6742203 | ||