siRNA Details
| Gene Symbol: Rmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025343.5 | NP_079619.4 | 12, 13 | 1517 | ||
| GenBank mRNA | AK009625.1 | - | 3, 4 | 305 | ||
| AK075603.1 | - | 12, 13 | 1500 | |||
| AK163523.1 | - | 753 | ||||
| AK212813.1 | - | 1, 2 | 18 | |||
| AK218009.1 | - | 1, 2 | 40 | |||
| BC019572.1 | - | 9, 10 | 762 | |||
| BC027299.1 | - | 12, 13 | 1465 | 3987394 | ||
| BC036150.1 | - | 11, 12 | 1437 | 4977394 | ||

Rmnd1
Genomic Map