siRNA Details
| Gene Symbol: Rmnd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025346.1 | NP_079622.1 | 7 | 1162 | ||
| GenBank mRNA | AK005344.1 | - | 4 | 541 | ||
| AK075599.1 | - | 7 | 1162 | |||
| AK179668.1 | - | 1 | 24 | |||
| BC016075.1 | - | 7 | 1016 | 4485143 | ||

Rmnd5b
Genomic Map