siRNA Details
| Gene Symbol: Rmnd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025346.1 | NP_079622.1 | 10 | 1530 | ||
| GenBank mRNA | AK005344.1 | - | 7 | 910 | ||
| AK075599.1 | - | 10 | 1530 | |||
| BC016075.1 | - | 10 | 1384 | 4485143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmnd5b| Gene Symbol: Rmnd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025346.1 | NP_079622.1 | 10 | 1530 | ||
| GenBank mRNA | AK005344.1 | - | 7 | 910 | ||
| AK075599.1 | - | 10 | 1530 | |||
| BC016075.1 | - | 10 | 1384 | 4485143 | ||