siRNA Details
| Gene Symbol: TRAFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143906.1 | NP_001137378.1 | 3 | 726 | ||
| NM_006700.2 | NP_006691.1 | 3 | 195 | |||
| GenBank mRNA | AB007447.1 | - | 3 | 163 | ||
| AK122620.1 | - | 3 | 711 | |||
| AK291331.1 | - | 3 | 180 | |||
| AK295474.1 | - | 3 | 180 | |||
| AK309916.1 | - | 3 | 202 | |||
| AK312601.1 | - | 3 | 195 | |||
| BC003553.2 | - | 2 | 121 | 3547039 | ||
| BG828961.1 | - | 3 | 699 | |||

TRAFD1
Genomic Map