siRNA Details
| Gene Symbol: Chchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025366.3 | NP_079642.1 | 1 | 160 | ||
| GenBank mRNA | AK003263.1 | - | 1 | 119 | ||
| AK010311.1 | - | 1 | 103 | |||
| AK011261.1 | - | 1 | 118 | |||
| AK027907.1 | - | 1 | 130 | |||
| AV460497.1 | - | 1 | 160 | |||
| BC049582.1 | - | 129 | 6772118 | |||
| BC052720.1 | - | 1 | 139 | 6818266 | ||
| BC064310.1 | - | 1 | 139 | |||
| BC092216.1 | - | 1 | 127 | 30609530 | ||

Chchd1
Genomic Map