siRNA Details
| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025370.2 | NP_079646.1 | 6 | 836 | ||
| GenBank mRNA | AK003782.1 | - | 6 | 837 | ||
| AK006589.1 | - | 6 | 691 | |||
| AK145073.1 | - | 834 | ||||
| AK145867.1 | - | 705 | ||||
| AK148379.1 | - | 815 | ||||
| AK168581.1 | - | 5 | 648 | |||
| AK168701.1 | - | 715 | ||||

Aaed1
Genomic Map