siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 4 | 683 | ||
| GenBank mRNA | AK038972.1 | - | 5 | 845 | ||
| AY364165.1 | - | 4 | 468 | |||
| AY846873.1 | - | 4 | 684 | |||
| AY846874.1 | - | 4 | 684 | |||
| BC019186.1 | - | 4 | 652 | |||
| BC042733.2 | - | 4 | 623 | |||
| BC043306.1 | - | 1 | 115 | |||
| BC044802.1 | - | 4 | 623 | |||
| BC054355.1 | - | 4 | 487 | |||
| BC054769.1 | - | 4 | 576 | 6415086 | ||

Tmem57
Genomic Map