siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 11 | 2584 | ||
| GenBank mRNA | AK014449.1 | - | 3 | 839 | ||
| AY364165.1 | - | 11 | 2369 | |||
| AY846873.1 | - | 11 | 2585 | |||
| BC043306.1 | - | 8 | 2016 | |||
| BC056944.1 | - | 3 | 2889 | |||

Tmem57
Genomic Map