siRNA Details
| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156566.1 | - | 3, 4 | 323 | ||
| AK170987.1 | - | 3, 4 | 331 | |||
| AK172538.1 | - | 3, 4 | 316 | |||
| BC037069.1 | - | 3, 4 | 345 | 5002765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Necap2| Gene Symbol: Necap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156566.1 | - | 3, 4 | 323 | ||
| AK170987.1 | - | 3, 4 | 331 | |||
| AK172538.1 | - | 3, 4 | 316 | |||
| BC037069.1 | - | 3, 4 | 345 | 5002765 | ||