siRNA Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164472.1 | NP_001157944.1 | 2 | 135 | ||
| NM_025391.2 | NP_079667.2 | 2 | 135 | |||
| GenBank mRNA | AK003739.1 | - | 2 | 135 | ||
| AK006057.1 | - | 2 | 114 | |||
| AK014290.1 | - | 2 | 94 | |||
| BE623972.1 | - | 2 | 106 | |||

Nip7
Genomic Map