siRNA Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025391.2 | NP_079667.2 | 4 | 378 | ||
| GenBank mRNA | AK003739.1 | - | 4 | 378 | ||
| AK006057.1 | - | 4 | 358 | |||
| AK014290.1 | - | 4 | 337 | |||
| BC003972.1 | - | 4 | 365 | 3488726 | ||
| BE623972.1 | - | 3 | 442 | |||

Nip7
Genomic Map