siRNA Details
| Gene Symbol: S100a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163525.2 | NP_001156997.1 | 4 | 732 | ||
| NM_001163526.2 | NP_001156998.1 | 3 | 568 | |||
| NM_025393.4 | NP_079669.2 | 4 | 715 | |||
| GenBank mRNA | AK003669.1 | - | 4 | 695 | ||
| AK132637.1 | - | 2 | 1332 | |||
| BC025607.1 | - | 3 | 714 | 5151924 | ||

S100a14
Genomic Map