siRNA Details
| Gene Symbol: Chchd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025395.3 | NP_079671.1 | 4 | 1051 | ||
| GenBank mRNA | AK003936.1 | - | 4 | 933 | ||
| AK013018.1 | - | 4 | 929 | |||
| BB792161.1 | - | 1 | 159 | |||
| BC027303.1 | - | 2 | 610 | |||
| W41426.1 | - | 1 | 87 | |||

Chchd5
Genomic Map