siRNA Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025400.3 | NP_079676.1 | 7 | 764 | ||
GenBank mRNA | AK003948.1 | - | 7 | 718 | ||
AK009262.1 | - | 6 | 494 | |||
AK020083.1 | - | 7 | 747 | |||
AK082469.1 | - | 7 | 704 | |||
AK148048.1 | - | 7 | 796 | |||
AK154593.1 | - | 6 | 895 | |||
BC011315.1 | - | 7 | 764 | 4013700 |
