siRNA Details
| Gene Symbol: Rps4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004068.1 | - | 1 | 560 | ||
| AK012382.1 | - | 1 | 559 | |||
| AK132160.1 | - | 1 | 560 | |||
| BC047994.1 | - | 1 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps4l| Gene Symbol: Rps4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004068.1 | - | 1 | 560 | ||
| AK012382.1 | - | 1 | 559 | |||
| AK132160.1 | - | 1 | 560 | |||
| BC047994.1 | - | 1 | 531 | |||