siRNA Details
| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025409.3 | NP_079685.1 | 3 | 592 | ||
| GenBank mRNA | AK004281.1 | - | 3 | 576 | ||
| BC021512.1 | - | 3 | 579 | 5351700 | ||
| CF951253.1 | - | 2 | 323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ier3ip1| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025409.3 | NP_079685.1 | 3 | 592 | ||
| GenBank mRNA | AK004281.1 | - | 3 | 576 | ||
| BC021512.1 | - | 3 | 579 | 5351700 | ||
| CF951253.1 | - | 2 | 323 | |||