siRNA Details
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025418.3 | NP_079694.2 | 4 | 401 | ||
GenBank mRNA | AK002611.1 | - | 4 | 400 | ||
AK003213.1 | - | 3 | 318 | |||
AK004323.1 | - | 4 | 473 | |||
AK009227.2 | - | 4 | 390 | |||
AK085518.1 | - | 1 | 636 | |||
BC026752.1 | - | 4 | 394 | 4034998 | ||
CX212626.1 | - | 4 | 402 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
