siRNA Details
| Gene Symbol: 1110059G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025419.4 | NP_079695.2 | 3 | 1040 | ||
| GenBank mRNA | AK083316.1 | - | 3 | 1013 | ||
| AK160617.1 | - | 3 | 1041 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110059G10Rik| Gene Symbol: 1110059G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025419.4 | NP_079695.2 | 3 | 1040 | ||
| GenBank mRNA | AK083316.1 | - | 3 | 1013 | ||
| AK160617.1 | - | 3 | 1041 | |||