siRNA Details
| Gene Symbol: 1110059E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025423.2 | NP_079699.1 | 4 | 919 | ||
| GenBank mRNA | AK004308.1 | - | 4 | 920 | ||
| BC023385.1 | - | 4 | 916 | 5253886 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110059E24Rik| Gene Symbol: 1110059E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025423.2 | NP_079699.1 | 4 | 919 | ||
| GenBank mRNA | AK004308.1 | - | 4 | 920 | ||
| BC023385.1 | - | 4 | 916 | 5253886 | ||