siRNA Details
| Gene Symbol: Nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025424.2 | NP_079700.1 | 2, 3 | 268 | ||
| GenBank mRNA | AB126218.1 | - | 2, 3 | 242 | ||
| AK002413.1 | - | 2, 3 | 269 | |||
| AK004329.1 | - | 2, 3 | 257 | |||
| AK009304.1 | - | 2, 3 | 259 | |||
| AK020403.1 | - | 2, 3 | 250 | |||
| AK155319.1 | - | 2, 3 | 267 | |||
| BC048464.1 | - | 2, 3 | 256 | 6596336 | ||

Nenf
Genomic Map