siRNA Details
| Gene Symbol: Rgcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025427.2 | NP_079703.2 | 3, 4 | 489 | ||
| GenBank mRNA | AF276981.1 | - | 3, 4 | 501 | ||
| AK004451.1 | - | 3, 4 | 490 | |||
| AK004705.1 | - | 3, 4 | 485 | |||
| BC049580.1 | - | 3, 4 | 485 | 6741281 | ||
| BC050935.1 | - | 3, 4 | 478 | 6818687 | ||
| CT010403.1 | - | 3, 4 | 351 | |||

Rgcc
Genomic Map