siRNA Details
| Gene Symbol: Tmem9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160145.1 | NP_001153617.1 | 6 | 934 | ||
| NM_001160146.1 | NP_001153618.1 | 6 | 830 | |||
| NM_025439.3 | NP_079715.1 | 6 | 799 | |||
| GenBank mRNA | AK005248.1 | - | 6 | 786 | ||
| AK134909.1 | - | 6 | 764 | |||
| AK154423.1 | - | 6 | 786 | |||
| AK186635.1 | - | 1 | 188 | |||
| AK186719.1 | - | 1 | 188 | |||
| BC016524.1 | - | 6 | 773 | 3594475 | ||
| BC020291.1 | - | 5 | 751 | |||
| BC048017.1 | - | 6 | 753 | |||

Tmem9
Genomic Map