siRNA Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025442.3 | NP_079718.1 | 2 | 159 | ||
| GenBank mRNA | AK003818.1 | - | 2 | 140 | ||
| AK005296.1 | - | 2 | 160 | |||
| AK006206.1 | - | 2 | 128 | |||
| AK011159.1 | - | 2 | 128 | |||
| AK030567.1 | - | 2 | 132 | |||
| AK161187.1 | - | 140 | ||||
| BC027160.1 | - | 2 | 119 | 4946430 | ||

Alg5
Genomic Map