siRNA Details
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025446.3 | NP_079722.1 | 6 | 927 | ||
| GenBank mRNA | AK008210.1 | - | 6 | 930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aig1| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025446.3 | NP_079722.1 | 6 | 927 | ||
| GenBank mRNA | AK008210.1 | - | 6 | 930 | ||