siRNA Details
| Gene Symbol: Ssr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025448.3 | NP_079724.1 | 5 | 735 | ||
| GenBank mRNA | AK005340.1 | - | 6 | 606 | ||
| AK007538.1 | - | 3 | 202 | |||
| AK007720.1 | - | 3 | 202 | |||
| AK010565.1 | - | 6 | 573 | |||
| AK012281.1 | - | 5 | 563 | |||
| AK020145.1 | - | 6 | 556 | |||
| AK050505.1 | - | 5 | 736 | |||
| AK084530.1 | - | 6 | 573 | |||
| AK214352.1 | - | 3 | 283 | |||
| AK217251.1 | - | 2 | 118 | |||
| BC010214.1 | - | 5 | 719 | 3497894 | ||

Ssr2
Genomic Map