siRNA Details
| Gene Symbol: Ssr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025448.3 | NP_079724.1 | 5 | 1003 | ||
| GenBank mRNA | AK005340.1 | - | 6 | 874 | ||
| AK007538.1 | - | 3 | 470 | |||
| AK007720.1 | - | 3 | 470 | |||
| AK010565.1 | - | 6 | 841 | |||
| AK012281.1 | - | 5 | 831 | |||
| AK020145.1 | - | 6 | 824 | |||
| AK050505.1 | - | 5 | 1004 | |||
| AK084530.1 | - | 6 | 841 | |||
| AK217251.1 | - | 2 | 386 | |||
| BC010214.1 | - | 5 | 987 | 3497894 | ||

Ssr2
Genomic Map