siRNA Details
Gene Symbol: Tm4sf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025453.3 | NP_079729.1 | 4 | 1314 | ||
GenBank mRNA | AK007532.1 | - | 4 | 1316 | ||
AK017195.1 | - | 4 | 1315 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tm4sf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025453.3 | NP_079729.1 | 4 | 1314 | ||
GenBank mRNA | AK007532.1 | - | 4 | 1316 | ||
AK017195.1 | - | 4 | 1315 |