siRNA Details
| Gene Symbol: Ece2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025462.2 | NP_079738.2 | 2 | 286 | ||
| NM_177940.1 | NP_808809.1 | 2 | 286 | |||
| NM_177941.1 | NP_808810.1 | 2 | 286 | |||
| GenBank mRNA | AF489569.1 | - | 2 | 105 | ||
| AF489570.1 | - | 2 | 105 | |||
| AK007407.1 | - | 2 | 295 | |||
| AK007791.1 | - | 2 | 288 | |||
| BC115541.2 | - | 2 | 288 | |||
| BC115542.1 | - | 2 | 288 | |||

Ece2
Genomic Map